Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenylmercuric Acetate 98%, Thermo Scientific™

Catalog No. AC130721000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC130721000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC130721000 Supplier Thermo Scientific Chemicals Supplier No. 130721000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-38-4
Molecular Formula C8H8HgO2
Molecular Weight (g/mol) 336.742
MDL Number MFCD00008691
InChI Key XEBWQGVWTUSTLN-UHFFFAOYSA-M
Synonym phenylmercuric acetate, phenylmercury acetate, acetoxyphenylmercury, anticon, liquiphene, dyanacide, gallotox, hostaquik, kwiksan, meracen
PubChem CID 16682730
ChEBI CHEBI:27684
IUPAC Name acetyloxy(phenyl)mercury
SMILES CC(=O)O[Hg]C1=CC=CC=C1

Specifications

Melting Point 148°C to 153°C
Color White to Cream
Infrared Spectrum Authentic
Assay Percent Range 98%
Linear Formula CH3CO2HgC6H5
Quantity 100 g
Fieser 06,458
Merck Index 15, 7411
Formula Weight 336.73
Percent Purity ≥97.5%
Physical Form Crystalline Powder
Chemical Name or Material Phenylmercuric acetate, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.