CAS RN 62-55-5
Thioacetamide TS, 4% (w/v), Ricca Chemical
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide, 99+%, ACS reagent
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide, ACS, 99% min
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide, 98%
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide, ≥99%, (ACS Reagent Grade), MP Biomedicals
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide, MP Biomedicals
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide, ACS Reagent Grade, Ricca Chemical
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide, 0.25% (w/v), Ricca Chemical
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide 98.0+%, TCI America™
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide, ACS Grade, LabChem™
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Thioacetamide, ACS, ≥99.0%, Solstice
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
Sigma Aldrich Thioacetamide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich Thioacetamide
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Thioacetamide, Crystal, BAKER ANALYZED™ Reagent, J.T. Baker™
CAS: 62-55-5 Molecular Formula: C2H5NS Molecular Weight (g/mol): 75.13 MDL Number: MFCD00008070 InChI Key: YUKQRDCYNOVPGJ-UHFFFAOYSA-N Synonym: thioacetamide,acetothioamide,thiacetamide,acetamide, thio,thioacetimidic acid,acetimidic acid, thio,usaf cb-21,usaf ek-1719,rcra waste number u218,acetic acid, thiono-, amide PubChem CID: 2723949 ChEBI: CHEBI:32497 IUPAC Name: ethanethioamide SMILES: CC(N)=S
- 1
- 2