Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thioacetamide, ACS, ≥99.0%, Solstice

Catalog No. 60047714 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Glass bottle
Poly bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-714 50 g Poly bottle
60-047-713 250 g Poly bottle
60-047-712 1 kg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-047-714 Supplier Solstice Supplier No. 8845050G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent, for the precipitation (of heavy metals),≥99.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62-55-5
Molecular Formula C2H5NS
Molecular Weight (g/mol) 75.13
MDL Number MFCD00008070
InChI Key YUKQRDCYNOVPGJ-UHFFFAOYSA-N
Synonym thioacetamide, acetothioamide, thiacetamide, acetamide, thio, thioacetimidic acid, acetimidic acid, thio, usaf cb-21, usaf ek-1719, rcra waste number u218, acetic acid, thiono-, amide
PubChem CID 2723949
ChEBI CHEBI:32497
IUPAC Name ethanethioamide
SMILES CC(N)=S

Specifications

Melting Point 108°C to 112°C
Packaging Poly bottle
Linear Formula CH3CSNH2
Quantity 50 g
Beilstein 506006
Formula Weight 75.13g/mol
Percent Purity ≥99.0%
Chemical Name or Material Thioacetamide
Hazard Statement P201-P273-P305 + P351 + P338-P308 + P313
RTECSNumber 200-541-4
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.