accessibility menu, dialog, popup

UserName

Viewing profile as user:
OSO

CAS RN 620-32-6

IUPAC Name: (phenylmethanesulfonylmethyl)benzene
Synonyms: dibenzylsulfone
Molecular Weight (g/mol): 246.32
Molecular Formula: C14H14O2S
InChi Key: AWHNUHMUCGRKRA-UHFFFAOYSA-N
SMILES: O=S(=O)(CC1=CC=CC=C1)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)