accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOClClNO

CAS RN 620-45-1

IUPAC Name: sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
Synonyms: sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzenolate
Molecular Weight (g/mol): 290.07
Molecular Formula: C12H6Cl2NNaO2
InChi Key: FHLDWQLHDYCXKI-UHFFFAOYSA-M
SMILES: [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1
Quantity 
  • (5)
  • (1)
  • (2)
  • (4)
Physical Form 
  • (5)
  • (1)
  • (1)
Grade 
  • (2)
  • (1)
  • (1)
Color 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (4)
  • (5)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)