Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloroindophenol sodium salt, MP Biomedicals™

Catalog No. ICN15011825 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15011825 25 g
ICN15011810 10 g
ICN15011801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15011825 Supplier MP Biomedicals, Inc Supplier No. 0215011825
Add to Cart
Add to Cart

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, 2,6-dichloroindophenol sodium, unii-kad7q8xo1y, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
PubChem CID 23697355
ChEBI CHEBI:948
IUPAC Name sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Color Blue-Green
Quantity 25 g
Formula Weight 290.08
Physical Form Crystalline Powder
Chemical Name or Material 2, 6-Dichloroindophenol sodium salt
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.