Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,6-Dichloroindophenol Sodium Salt , MP Biomedicals, LLC

Catalog No. MP215011801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MP215011801 1 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP215011801 Supplier MP Biomedicals, Inc Supplier No. 0215011801
Add to Cart
Add to Cart

2,6-dichlorophenolindophenol sodium salt ( DCPIP ) is a blue chemical compound used as a redox dye. In Oxidized form DCPIP is blue; whereas in reduced form DCPIP is colorless.

  • 2,6-dichlorophenolindophenol sodium salt is a redox dye commonly used as a monitor of the light reactions in photosynthesis because it is an electron acceptor that is blue when oxidized and colorless when reduced
  • It is part of the Hill reagents family
  • DCPIP can also be used as an indicator for Vitamin C
  • If vitamin C, which is a good reducing agent, is present, the blue dye, which turns pink in acid conditions, is reduced to a colorless compound by ascorbic acid
  • The rate of photosynthesis can be measured by the rate at which this dye changes color (is reduced) when exposed to light in a photosynthetic system
  • This reaction is reversible, as colorless DCPIP can be reoxidized to blue 2,6-dichlorophenolindophenol (DCPIP) is used as a substitute for NADP+
  • The dye changes color when it is reduced, due to its chemical structure
  • The rate of photosynthesis light-dependent reaction can be measured with this property of DCPIP because one of the stages of the light reaction is an electron transport chain that normally ends with the reduction of NADP+
  • When DCPIP is present, it also gets reduced by the light reaction
  • The amount of DCPIP reduced can be found by measuring the solution's light transmittance with a spectrophotometer
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-45-1
Molecular Formula C12H6Cl2NNaO2
Molecular Weight (g/mol) 290.07
MDL Number MFCD00150014
InChI Key FHLDWQLHDYCXKI-UHFFFAOYSA-M
Synonym 2,6-dichloroindophenol sodium salt, 2,6-dichlorophenolindophenol sodium salt, tillman's reagent, tillman's reagenz, dichlorphenol-indophenolnatrium, unii-kad7q8xo1y, 2,6-dichloroindophenol sodium, sodium 2,6-dichloroindophenol, sodium 2,6-dichloroindophenolate, 2,6-dichloroindophenol, sodium salt
PubChem CID 23697355
ChEBI CHEBI:948
IUPAC Name sodium 2,6-dichloro-4-[(4-oxocyclohexa-2,5-dien-1-ylidene)amino]benzen-1-olate
SMILES [Na+].[O-]C1=C(Cl)C=C(C=C1Cl)N=C1C=CC(=O)C=C1

Specifications

Color Green
Packaging Glass
Quantity 1 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.