accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 621-52-3

IUPAC Name: 1-ethoxy-3-nitrobenzene
Synonyms: 3-ethoxynitrobenzene
Molecular Weight (g/mol): 167.16
Molecular Formula: C8H9NO3
InChi Key: LFOLBPDHVGDKGJ-UHFFFAOYSA-N
SMILES: CCOC1=CC=CC(=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results