accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 621-77-2

IUPAC Name: tripentylamine
Synonyms: 1-pentanamine, N,N-dipentyl-
Molecular Weight (g/mol): 227.44
Molecular Formula: C15H33N
InChi Key: OOHAUGDGCWURIT-UHFFFAOYSA-N
SMILES: CCCCCN(CCCCC)CCCCC
Molecular Weight (g/mol) 
  • (2)
  • (4)
Percent Purity 
  • (1)
  • (2)
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (4)