Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic Acid, Disodiumsalt Dihydrate, Reagent ACS, +99%, Thermo Scientific™

Catalog No. AC422151000
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC422151000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC422151000 Supplier Thermo Scientific Chemicals Supplier No. 422151000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-77-2
Molecular Formula C15H33N
Molecular Weight (g/mol) 227.436
InChI Key OOHAUGDGCWURIT-UHFFFAOYSA-N
Synonym triamylamine, tripentylamine, tri-n-pentylamine, 1-pentanamine, n,n-dipentyl, tri-n-amylamine, ccris 4877, tripentyl amine, 1-pentanamine,n-dipentyl, acmc-209n0e, 1-pentanamine,n,n-dipentyl
PubChem CID 12133
IUPAC Name N,N-dipentylpentan-1-amine
SMILES CCCCCN(CCCCC)CCCCC

Specifications

Color Yellow
Density 0.782g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Refractive Index 1.4360 to 1.4380 (20°C, 589nm)
Linear Formula (CH3(CH2)4)3N
Quantity 100 g
Formula Weight 227.43
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Tripentylamine, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.