accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 622-61-7

IUPAC Name: 1-chloro-4-ethoxybenzene
Synonyms: benzene, 1-chloro-4-ethoxy-
Molecular Weight (g/mol): 156.61
Molecular Formula: C8H9ClO
InChi Key: IXLSVQMYQRAMEW-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results