accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 622-93-5

IUPAC Name: 3-(diethylamino)propan-1-ol
Synonyms: 1-propanol, 3-(diethylamino)-
Molecular Weight (g/mol): 131.22
Molecular Formula: C7H17NO
InChi Key: WKCYFSZDBICRKL-UHFFFAOYSA-N
SMILES: CCN(CC)CCCO
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)