accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOFFFOOFFF

CAS RN 62240-27-1

IUPAC Name: 1,2-bis(2,2,2-trifluoroethyl) benzene-1,2-dicarboxylate
Synonyms: 1,2-bis(2,2,2-trifluoroethyl) phthalate
Molecular Weight (g/mol): 330.18
Molecular Formula: C12H8F6O4
InChi Key: PSRBRNHUQJKQHV-UHFFFAOYSA-N
SMILES: FC(F)(F)COC(=O)C1=CC=CC=C1C(=O)OCC(F)(F)F
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results