accessibility menu, dialog, popup

UserName

Viewing profile as user:
NOOHNH2

CAS RN 62247-21-6

IUPAC Name: 3-amino-3-(pyridin-3-yl)propanoic acid
Synonyms: 3-amino-3-(pyridin-3-yl)propanoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: QOTCEJINJFHMLO-UHFFFAOYNA-N
SMILES: NC(CC(O)=O)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results