accessibility menu, dialog, popup

UserName

Viewing profile as user:
FFFOSOOFFF

CAS RN 6226-25-1

IUPAC Name: 2,2,2-trifluoroethyl trifluoromethanesulfonate
Synonyms: 2,2,2-trifluoroethyl trifluoromethanesulfonate
Molecular Weight (g/mol): 232.1
Molecular Formula: C3H2F6O3S
InChi Key: RTMMSCJWQYWMNK-UHFFFAOYSA-N
SMILES: FC(F)(F)COS(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (1)
  • (4)
  • (3)
Quantity 
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (2)
  • (3)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (5)
  • (1)
  • (4)
  • (3)
  • (1)
  • (3)
  • (4)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (4)