accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrHNOS

CAS RN 62266-82-4

IUPAC Name: 6-bromo-2,3-dihydro-1,3-benzothiazol-2-one
Synonyms: 6-bromo-3H-1,3-benzothiazol-2-one
Molecular Weight (g/mol): 230.08
Molecular Formula: C7H4BrNOS
InChi Key: HECJMTPEVWQFCY-UHFFFAOYSA-N
SMILES: BrC1=CC=C2NC(=O)SC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results