accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 623-42-7

IUPAC Name: methyl butanoate
Synonyms: methyl butyrate
Molecular Weight (g/mol): 102.13
Molecular Formula: C5H10O2
InChi Key: UUIQMZJEGPQKFD-UHFFFAOYSA-N
SMILES: CCCC(=O)OC
Percent Purity 
  • (1)
  • (2)
  • (3)
  • (6)
Boiling Point 
  • (1)
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (4)
  • (1)
  • (2)
  • (3)
  • (6)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (6)
  • (2)