accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 623-46-1

IUPAC Name: 1,2-dichloro-1-ethoxyethane
Synonyms: ethane, 1,2-dichloro-1-ethoxy-
Molecular Weight (g/mol): 143.01
Molecular Formula: C4H8Cl2O
InChi Key: NNBUKAPOVBEMNI-UHFFFAOYNA-N
SMILES: CCOC(Cl)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results