Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Dichloroethyl Ethyl Ether 93.0+%, TCI America™
SDP

Catalog No. D035525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D035525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D035525G Supplier TCI America Supplier No. D035525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 623-46-1
Molecular Formula C4H8Cl2O
Molecular Weight (g/mol) 143.01
MDL Number MFCD00041730
InChI Key NNBUKAPOVBEMNI-UHFFFAOYNA-N
Synonym 1,2-Dichlorodiethyl Ether
PubChem CID 79077
IUPAC Name 1,2-dichloro-1-ethoxyethane
SMILES CCOC(Cl)CCl

Specifications

Color Yellow
Boiling Point 145°C
Quantity 25 g
UN Number 3271
Formula Weight 143.01
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2-Dichloroethyl Ethyl Ether
RTECSNumber KI0780000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.