accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCl

CAS RN 623-71-2

IUPAC Name: ethyl 3-chloropropanoate
Synonyms: ethyl 3-chloropropanoate
Molecular Weight (g/mol): 136.58
Molecular Formula: C5H9ClO2
InChi Key: ZCLGVXACCAZJOX-UHFFFAOYSA-N
SMILES: CCOC(=O)CCCl
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)