accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COSOOCH3

CAS RN 623-98-3

IUPAC Name: dipropyl sulfite
Synonyms: dipropyl sulfite
Molecular Weight (g/mol): 166.24
Molecular Formula: C6H14O3S
InChi Key: MAIQPVFXODAAIG-UHFFFAOYSA-N
SMILES: CCCOS(=O)OCCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results