accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6232-57-1

IUPAC Name: 4-[2-(4-aminophenyl)diazen-1-yl]-2-methoxy-5-methylaniline
Synonyms: 4-[2-(4-aminophenyl)diazen-1-yl]-2-methoxy-5-methylaniline
Molecular Weight (g/mol): 256.31
Molecular Formula: C14H16N4O
InChi Key: DBENZRUFCCLWPP-UHFFFAOYSA-N
SMILES: COC1=CC(N=NC2=CC=C(N)C=C2)=C(C)C=C1N
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results