accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOPOOH3COH3C

CAS RN 62327-21-3

IUPAC Name: tert-butyl 2-(dimethoxyphosphoryl)acetate
Synonyms: tert-butyl 2-(dimethoxyphosphoryl)acetate
Molecular Weight (g/mol): 224.19
Molecular Formula: C8H17O5P
InChi Key: SAZYDWOWLRDDRQ-UHFFFAOYSA-N
SMILES: COP(=O)(CC(=O)OC(C)(C)C)OC
Boiling Point 
  • (1)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)