accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 625-06-9

IUPAC Name: 2,4-dimethylpentan-2-ol
Synonyms: 2,4-dimethyl-2-pentanol
Molecular Weight (g/mol): 116.2
Molecular Formula: C7H16O
InChi Key: FMLSQAUAAGVTJO-UHFFFAOYSA-N
SMILES: CC(C)CC(C)(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results