accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOCH3

CAS RN 626-29-9

IUPAC Name: tetradecanoyl tetradecanoate
Synonyms: tetradecanoic acid, anhydride
Molecular Weight (g/mol): 438.74
Molecular Formula: C28H54O3
InChi Key: RCRYHUPTBJZEQS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCC(=O)OC(=O)CCCCCCCCCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)