accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SNNHSS

CAS RN 6264-40-0

IUPAC Name: 5-(methylsulfanyl)-2,3-dihydro-1,3,4-thiadiazole-2-thione
Synonyms: 5-(methylsulfanyl)-3H-1,3,4-thiadiazole-2-thione
Molecular Weight (g/mol): 164.26
Molecular Formula: C3H4N2S3
InChi Key: CFWGYKRJMYXYND-UHFFFAOYSA-N
SMILES: CSC1=NNC(=S)S1
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)