accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 627034-85-9

IUPAC Name: pentasodium 4-{3-[(2,4-disulfonatophenyl)carbamoyl]-5-[({3-[(2,4-disulfophenyl)carbamoyl]-5-[(2-sulfo-4-sulfonatophenyl)carbamoyl]phenyl}carbamoyl)amino]benzamido}benzene-1,3-disulfonate
Synonyms: pentasodium 4-{3-[(2,4-disulfonatophenyl)carbamoyl]-5-[({3-[(2,4-disulfophenyl)carbamoyl]-5-[(2-sulfo-4-sulfonatophenyl)carbamoyl]phenyl}carbamoyl)amino]benzamido}benzene-1,3-disulfonate
Molecular Weight (g/mol): 1439.11
Molecular Formula: C41H27N6Na5O29S8
InChi Key: PHCBPKWKKHYRSA-UHFFFAOYSA-I
SMILES: [Na+].[Na+].[Na+].[Na+].[Na+].OS(=O)(=O)C1=CC(=C(NC(=O)C2=CC(NC(=O)NC3=CC(=CC(=C3)C(=O)NC3=C(C=C(C=C3)S([O-])(=O)=O)S([O-])(=O)=O)C(=O)NC3=C(C=C(C=C3)S([O-])(=O)=O)S([O-])(=O)=O)=CC(=C2)C(=O)NC2=C(C=C(C=C2)S([O-])(=O)=O)S(O)(=O)=O)C=C1)S(O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results