accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62720-29-0

IUPAC Name: 1,3-dibromo-5-fluoro-2-iodobenzene
Synonyms: 1,3-dibromo-5-fluoro-2-iodobenzene
Molecular Weight (g/mol): 379.79
Molecular Formula: C6H2Br2FI
InChi Key: LIWKANDEJFABTQ-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(I)C(Br)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results