accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 62733-99-7

IUPAC Name: methyl 3-hydroxypyridine-2-carboxylate
Synonyms: methyl 3-hydroxypyridine-2-carboxylate
Molecular Weight (g/mol): 153.14
Molecular Formula: C7H7NO3
InChi Key: MHKKUZDJUGIOBC-UHFFFAOYSA-N
SMILES: COC(=O)C1=C(O)C=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results