accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3C(R)CH3OONH2(E)

CAS RN 62742-50-1

IUPAC Name: (2R)-2-methylbutyl (2E)-3-(4-aminophenyl)prop-2-enoate
Synonyms: (2R)-2-methylbutyl (2E)-3-(4-aminophenyl)prop-2-enoate
Molecular Weight (g/mol): 233.31
Molecular Formula: C14H19NO2
InChi Key: OUZTVIGSXSIPAZ-OALRPVAKSA-N
SMILES: CC[C@@H](C)COC(=O)\C=C\C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results