accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6278-73-5

IUPAC Name: 4-(1,3-benzothiazol-2-yl)aniline
Synonyms: 4-(1,3-benzothiazol-2-yl)aniline
Molecular Weight (g/mol): 226.3
Molecular Formula: C13H10N2S
InChi Key: WKRCOZSCENDENK-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=NC2=CC=CC=C2S1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results