accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONNBOHHO

CAS RN 628692-15-9

IUPAC Name: (2-methoxypyrimidin-5-yl)boronic acid
Synonyms: 2-methoxypyrimidin-5-ylboronic acid
Molecular Weight (g/mol): 153.93
Molecular Formula: C5H7BN2O3
InChi Key: YPWAJLGHACDYQS-UHFFFAOYSA-N
SMILES: COC1=NC=C(C=N1)B(O)O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results