accessibility menu, dialog, popup

UserName

Viewing profile as user:
HOOH

CAS RN 629-11-8

IUPAC Name: hexane-1,6-diol
Synonyms: hexane-1,6-diol α,ω-hexanediol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: XXMIOPMDWAUFGU-UHFFFAOYSA-N
SMILES: OCCCCCCO
Molecular Weight (g/mol) 
  • (4)
  • (6)
  • (3)
Boiling Point 
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
Percent Purity 
  • (2)
  • (9)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (4)
  • (1)
  • (2)
  • (6)
  • (1)
  • (1)
  • (4)
  • (6)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (9)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)