CAS RN 629-64-1
CAS RN 629-64-1
IUPAC Name:
1-(heptyloxy)heptane
Synonyms:
heptane, 1,1'-oxybis-
Molecular Weight (g/mol):
214.39
Molecular Formula:
C14H30O
InChi Key:
UJEGHEMJVNQWOJ-UHFFFAOYSA-N
SMILES:
CCCCCCCOCCCCCCC
1
–
1
of
1
results
1
Heptyl Ether 98.0+%, TCI America™
CAS: 629-64-1 Molecular Formula: C14H30O Molecular Weight (g/mol): 214.39 MDL Number: MFCD00015288 InChI Key: UJEGHEMJVNQWOJ-UHFFFAOYSA-N Synonym: Diheptyl Ether PubChem CID: 12392 IUPAC Name: 1-(heptyloxy)heptane SMILES: CCCCCCCOCCCCCCC