Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptyl Ether 98.0+%, TCI America™
SDP

Catalog No. H036425ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H036425ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H036425ML Supplier TCI America Supplier No. H036425ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 629-64-1
Molecular Formula C14H30O
Molecular Weight (g/mol) 214.39
MDL Number MFCD00015288
InChI Key UJEGHEMJVNQWOJ-UHFFFAOYSA-N
Synonym Diheptyl Ether
PubChem CID 12392
IUPAC Name 1-(heptyloxy)heptane
SMILES CCCCCCCOCCCCCCC

Specifications

Color Colorless
Boiling Point 262°C
Quantity 25 mL
Formula Weight 214.39
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptyl Ether
RTECSNumber MK1390500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.