accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 629-98-1

IUPAC Name: (13Z)-docos-13-en-1-ol
Synonyms: (13Z)-docos-13-en-1-ol
Molecular Weight (g/mol): 324.59
Molecular Formula: C22H44O
InChi Key: CFOQKXQWGLAKSK-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCCCCCO
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results