accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3

CAS RN 6297-41-2

IUPAC Name: butyl 2-methylpentanoate
Synonyms: butyl isohexanoate
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: FMVUGLMJUHFPSJ-UHFFFAOYNA-N
SMILES: CCCCOC(=O)C(C)CCC
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results