accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3

CAS RN 630-02-4

IUPAC Name: octacosane
Synonyms: octacosane N-octacosane ccris 680
Molecular Weight (g/mol): 394.77
Molecular Formula: C28H58
InChi Key: ZYURHZPYMFLWSH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCCCCCCCC
Molecular Weight (g/mol) 
  • (1)
  • (11)
Quantity 
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (1)
  • (4)
  • (4)
Physical Form 
  • (3)
  • (1)
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (7)
  • (1)
  • (11)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (4)
  • (4)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)