Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

n-Octacosane, ≥99%, MP Biomedicals™

Catalog No. ICN15596425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
ICN15596425 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15596425 Supplier MP Biomedicals, Inc Supplier No. 0215596425

missing translation for 'validMainframeMsg'

Specifications

CAS 630-02-4
Color White
Assay Percent Range ≥99%
Molecular Formula C28H58
Quantity 25 g
MDL Number MFCD00009355
Synonym n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
InChI Key ZYURHZPYMFLWSH-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCC
IUPAC Name octacosane
Molecular Weight (g/mol) 394.77
PubChem CID 12408
ChEBI CHEBI:32943
Formula Weight 394.8
Physical Form Solid
Chemical Name or Material n-Octacosane
Show More Show Less
EINECSNumber 211-125-7
Recommended Storage Store at room temperature (15° to 30°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.