accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OSCH3

CAS RN 63012-97-5

IUPAC Name: 2-methyl-3-(methylsulfanyl)furan
Synonyms: 2-methyl-3-(methylsulfanyl)furan
Molecular Weight (g/mol): 128.19
Molecular Formula: C6H8OS
InChi Key: OQVAOEIMSKZGAL-UHFFFAOYSA-N
SMILES: CSC1=C(C)OC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results