accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 6303-21-5

IUPAC Name: phosphinic acid
Synonyms: phosphinic acid|hypophosphorous|hypophosphorous acid|H2PO(OH)|hydrophosphorous acid|HPA
Molecular Weight (g/mol): 66
Molecular Formula: H3O2P
InChi Key: ACVYVLVWPXVTIT-UHFFFAOYSA-N
SMILES: O[PH2]=O
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results