accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 63056-20-2

IUPAC Name: 6-phenylpyridine-3-carbaldehyde
Synonyms: 6-phenylpyridine-3-carbaldehyde
Molecular Weight (g/mol): 183.21
Molecular Formula: C12H9NO
InChi Key: HWDMDOSKSDLXEH-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(N=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results