accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3BrONH2NHN

CAS RN 6307-35-3

IUPAC Name: 2-amino-5-bromo-6-methyl-1,4-dihydropyrimidin-4-one
Synonyms: 2-amino-5-bromo-6-methyl-1H-pyrimidin-4-one
Molecular Weight (g/mol): 204.03
Molecular Formula: C5H6BrN3O
InChi Key: ADLWOFHKMXUDKF-UHFFFAOYSA-N
SMILES: CC1=C(Br)C(=O)N=C(N)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results