accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH2NCH3H2CCH3H2C

CAS RN 6321-40-0

IUPAC Name: tris(2-methylprop-2-en-1-yl)amine
Synonyms: tris(2-methylprop-2-en-1-yl)amine
Molecular Weight (g/mol): 179.31
Molecular Formula: C12H21N
InChi Key: FXBJYRVIFGLPBC-UHFFFAOYSA-N
SMILES: CC(=C)CN(CC(C)=C)CC(C)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results