accessibility menu, dialog, popup

UserName

Viewing profile as user:
NSeNBrBr

CAS RN 63224-42-0

IUPAC Name: 4,7-dibromo-2,1,3-benzoselenadiazole
Synonyms: 4,7-dibromo-2,1,3-benzoselenadiazole
Molecular Weight (g/mol): 340.88
Molecular Formula: C6H2Br2N2Se
InChi Key: MVYRQFKGUCDJAB-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)C2=N[Se]N=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results