CAS RN 63231-69-6
Molecular sieves, 13X, 1.6-2.5 mm (0.063-0.098in) beads
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular sieves, 13X, 3-5 mm (0.12-0.20in) beads
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular sieves, 13X, 1-2mm (0.04-0.08in) dia. pellets
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular sieves, 13X, powder
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular sieves 13X, 4 to 8 mesh
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular sieves, 13X, 3-4mm (0.12-0.16in) dia. pellets
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular sieves 13X, 8 to 12 mesh
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular Sieves, 13X, Solstice
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular Sieves, 13X, 8 to 12mesh, Solstice
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular Sieves, 13X, Solstice
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular Sieves, 13X, Pellets, Dia.: 1.6mm, Solstice
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular Sieves, 13X, Pellets, Dia.: 3.2mm, Solstice
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X
Molecular Sieves, 13X, Solstice
CAS: 63231-69-6 MDL Number: MFCD00131613 IUPAC Name: Molecular sieves 13X