Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Molecular Sieves, 13X, Pellets, Dia.: 3.2mm, Solstice

Catalog No. 60046255 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Drum with inliner
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-255 5 kg Drum with inliner
60-046-254 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-046-255 Supplier Solstice Supplier No. 3343595KG

missing translation for 'validMainframeMsg'

Pellets, 3.2 mm

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63231-69-6
MDL Number MFCD00131613
IUPAC Name Molecular sieves 13X

Specifications

Quantity 5 kg
Particle Size Dia.: 3.2mm
Packaging Drum with inliner
Physical Form Pellets
Linear Formula Na86[AlO2)86(SiO2)106] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular sieves, 13X
Hazard Statement P261-P305 + P351 + P338
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.