accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH

CAS RN 6329-61-9

IUPAC Name: decahydroisoquinoline
Synonyms: decahydroisoquinoline
Molecular Weight (g/mol): 139.24
Molecular Formula: C9H17N
InChi Key: NENLYAQPNATJSU-UHFFFAOYNA-N
SMILES: C1CCC2CNCCC2C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)