accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NSONHClH2NCH3

CAS RN 633283-39-3

IUPAC Name: 3-amino-N-[(4-chlorophenyl)methyl]-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide
Synonyms: 3-amino-N-[(4-chlorophenyl)methyl]-4,6-dimethylthieno[2,3-b]pyridine-2-carboxamide
Molecular Weight (g/mol): 345.85
Molecular Formula: C17H16ClN3OS
InChi Key: FPRULFHDSFKYBV-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C2C(N)=C(SC2=N1)C(=O)NCC1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results