accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2SOOS

CAS RN 6339-87-3

IUPAC Name: thiophene-2-sulfonamide
Synonyms: 2-thiophenesulfonamide sulfamido-2 thiophene
Molecular Weight (g/mol): 163.21
Molecular Formula: C4H5NO2S2
InChi Key: KTFDYVNEGTXQCV-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=CS1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results